Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropionic acid, 99%, Thermo Scientific™

Catalog No. AC161051000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC161051000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161051000 Supplier Thermo Scientific Chemicals Supplier No. 161051000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 600-05-5
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C3H4Br2O2
Linear Formula BrCH2CH(Br)CO2H
MDL Number MFCD00004212
Quantity 100g
Beilstein 02, 258
Synonym 2,3-dibromopropionic acid, propanoic acid, 2,3-dibromo, alpha,beta-dibromopropionic acid, propionic acid, 2,3-dibromo, .alpha.,.beta.-dibromopropionic acid, acmc-209mhg, propanoic acid,3-dibromo, propionic acid,3-dibromo, 2,3-dibromo-propionic acid, dsstox_cid_12832
InChI Key ZMYAKSMZTVWUJB-UHFFFAOYNA-N
SMILES OC(=O)C(Br)CBr
IUPAC Name 2,3-dibromopropanoic acid
Molecular Weight (g/mol) 231.87
PubChem CID 11746
Formula Weight 231.87
Percent Purity ≥98.5%
Chemical Name or Material 2, 3-Dibromopropionic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.