Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-1,4-naphthoquinone, 98%

Catalog No. AC113480250
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113480250 25 g Glass bottle
AC113481000 100 g Glass bottle
AC113482500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC113480250 Supplier Thermo Scientific Chemicals Supplier No. 113480250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 117-80-6 | C10H4Cl2O2 | 227.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-80-6
Molecular Formula C10H4Cl2O2
Molecular Weight (g/mol) 227.04
MDL Number MFCD00001677
InChI Key SVPKNMBRVBMTLB-UHFFFAOYSA-N
Synonym 2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone
PubChem CID 8342
SMILES ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 194.0°C to 197.0°C
Color Yellow
Boiling Point 275.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 07, 729
Merck Index 15, 3054
Solubility Information Solubility in water: 0.008g/L water. Other solubilities: soluble in xylene,47% in o-dichlorobenzene
IUPAC Name 2,3-dichloronaphthalene-1,4-dione
Formula Weight 227.05
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 3-Dichloro-1, 4-naphthoquinone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-210-5
RTECSNumber QL7525000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.