Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Dichloro-1-propanol

Catalog No. 362955ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-295-5ML 5 mL
36-295-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-295-5ML Supplier Sigma Aldrich Supplier No. 362955ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3-dichloro-1-propanol belongs to the group of chloropropanols
  • Inhibitory effects of 2,3-dichloro-1-propanol on t cell both in vivo and in vitro is reported
  • Improved enantioselective resolution of (r,s)-2,3-dichloro-1-propanol to (s)-2, 3-dichloro-1-propanol by whole cells of a recombinant escherichia coli in n-heptane-aqueous biphasic system is reported.2,3-dichloro-1-propanol may be employed as carbon and energy supplement for the growth of pseudomonas putida strain (mc4)

Specifications

CAS 616-23-9
Density 1.360 g/mL (at 20°C (literature))
Molecular Formula C3H6Cl2O
Linear Formula ClCH2CHClCH2OH
MDL Number MFCD00040446
Quantity 5 mL
Synonym Glycerol-alpha,beta-dichlorohydrin
Molecular Weight (g/mol) 128.99
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.