Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-dichloro-1-propanol-d5 | 1189730-34-4 | 98.0% | 134.02 g/mol | C3HD5Cl2O | 10 MG
SDP

Catalog No. 5000208308
Encompass_Preferred

2,3-dichloro-1-propanol-d5 is a deuterium-labeled analogue of 2,3-dichloro-1-propanol intended for use as an internal standard and tracer in analytical workflows such as mass spectrometry. It is supplied at high isotopic purity suitable for quantitative and qualitative analyses.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Purity 98.0%.
  • Chemical formula C3HD5Cl2O.
  • Molecular weight 134.02 g/mol.
  • Available in small-scale pack sizes for analytical use.

Catalog No. 50-002-08308 Supplier Medchemexpress LLC Supplier No. HYW087048S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-dichloro-1-propanol-d5 is a deuterium-labeled analogue of 2,3-dichloro-1-propanol intended for use as an internal standard and tracer in analytical workflows such as mass spectrometry. It is supplied at high isotopic purity suitable for quantitative and qualitative analyses.

  • Deuterium-labeled internal standard for mass spectrometry.
  • Purity 98.0%.
  • Chemical formula C3HD5Cl2O.
  • Molecular weight 134.02 g/mol.
  • Available in small-scale pack sizes for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.