Learn More
Medchemexpress LLC 2,3-dichloro-1-propanol-d5 | 1189730-34-4 | 98.0% | 134.02 g/mol | C3HD5Cl2O | 5 MG
SDP
Catalog No. 5000208529
Encompass_Preferred
2,3-Dichloro-1-propanol-d5 is a deuterium-labeled analogue of 2,3-dichloro-1-propanol supplied for research use. It functions as a tracer and internal standard for quantitative analyses and is characterized with a certificate of analysis and safety data sheet.
  • Deuterium-labeled tracer for analytical studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Purity 98.0%.
  • Available in small quantities, including 5 mg.
  • Store pure material at -20 °C; in solvent store at -80 °C or -20 °C.
Catalog No. 50-002-08529 Supplier Medchemexpress LLC Supplier No. HYW087048S5MG
2,3-Dichloro-1-propanol-d5 is a deuterium-labeled analogue of 2,3-dichloro-1-propanol supplied for research use. It functions as a tracer and internal standard for quantitative analyses and is characterized with a certificate of analysis and safety data sheet.
  • Deuterium-labeled tracer for analytical studies.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Purity 98.0%.
  • Available in small quantities, including 5 mg.
  • Store pure material at -20 °C; in solvent store at -80 °C or -20 °C.