Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-1-propene, 98%

Catalog No. AC113695000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113695000 500 mL Glass bottle
AC113691000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113695000 Supplier Thermo Scientific Chemicals Supplier No. 113695000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 78-88-6 | C3H4Cl2 | 110.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-88-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000943
InChI Key FALCMQXTWHPRIH-UHFFFAOYSA-N
Synonym 2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41
PubChem CID 6565
IUPAC Name 2,3-dichloroprop-1-ene
SMILES C=C(CCl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown-Pink to Colorless
Density 1.2000g/mL
Boiling Point 94°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4601 to 1.4621
Linear Formula H2C=C(Cl)CH2Cl
Quantity 500 mL
Beilstein 01, 199
Fieser 02,120; 08,158; 14,129
Specific Gravity 1.2
Formula Weight 110.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dichloro-1-propene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled. Harmful if swallowed. Harmful to aquatic life with long lasting effects. Suspected of causing genetic defects. Harmful in contact with skin. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
EINECSNumber 201-153-8
RTECSNumber UC8400000
TSCA TSCA
Recommended Storage may discolor to brown on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.