Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%

Catalog No. AAA1187918
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1187918 50 g
AAA1187909 10 g
AAA1187930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1187918 Supplier Thermo Scientific Chemicals Supplier No. A1187918
Only null left
Add to Cart

CAS: 84-58-2 | C8Cl2N2O2 | 227 g/mol

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone is used as a reagent for oxidative couplings and cyclization reactions and dehydrogenation of alcohols, phenols and steroid ketones. It is also used in the synthesis of 1,2-benzisoxazoles. It is a useful electron-transfer reagent for synthesis of quinolines from imines and alkynes or alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone is used as a reagent for oxidative couplings and cyclization reactions and dehydrogenation of alcohols, phenols and steroid ketones. It is also used in the synthesis of 1,2-benzisoxazoles. It is a useful electron-transfer reagent for synthesis of quinolines from imines and alkynes or alkenes.

Solubility
Soluble in benzene, methanol, acetic acid and dioxane.Slightly soluble in chloroform,, dichloromethane.

Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-58-2
Molecular Formula C8Cl2N2O2
Molecular Weight (g/mol) 227
MDL Number MFCD00001593
InChI Key HZNVUJQVZSTENZ-UHFFFAOYSA-N
Synonym 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
PubChem CID 6775
IUPAC Name 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N

Specifications

Melting Point 214°C to 218°C
Quantity 50 g
UN Number UN3439
Beilstein 747939
Sensitivity Moisture sensitive
Merck Index 14,3063
Solubility Information Soluble in benzene,methanol,acetic acid and dioxane.Slightly soluble in chloroform,,dichloromethane.
Formula Weight 227.01
Percent Purity ≥98%
Chemical Name or Material 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
Hazard Category H301-H318
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280-P301+P310-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 201-542-2
RTECSNumber GU4825000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.