Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Dichloro-5,6-Dicyano-P-Benzoquinone

Catalog No. D6040010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D6040010G 10 g
D60400100G 100 g
D604005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D6040010G Supplier Sigma Aldrich Supplier No. D6040010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,3-Dichloro-5,6-Dicyano-P-Benzoquinone

Specifications

CAS 644-97-3
Melting Point -51°C
Density 1.319 g/mL at 25°C
Boiling Point 222°C (759 mmHg, lit.)
Molecular Formula C6H5Cl2P
Linear Formula C6H5PCl2
Quantity 10 g
MDL Number MFCD00000528
Synonym 4,5-Dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile; DDQ
Molecular Weight (g/mol) 178.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.