Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-5-(trifluoromethyl)pyridine 95.0+%, TCI America™
SDP

Catalog No. D221425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D221425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D221425G Supplier TCI America Supplier No. D221425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorinated building block for organic synthesis, supplied at 95.0% purity.

Chemical Identifiers

CAS 69045-84-7
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD00042243
InChI Key ABNQGNFVSFKJGI-UHFFFAOYSA-N
Synonym 2,3-dichloro-5-trifluoromethyl pyridine, unii-5f2c8lx8ie, 2,3-dichloro-5-trifluoro methylpyridine, 2,3,5-dctf, 5f2c8lx8ie, 2,3-dichloro-5-trifluoromethyl-pyridine, pyridine, 2,3-dichloro-5-trifluoromethyl, pubchem9237
PubChem CID 112234
IUPAC Name 2,3-dichloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CN=C(Cl)C(Cl)=C1

Specifications

Color Colorless
Boiling Point 80°C
Quantity 25 g
UN Number 3082
Formula Weight 215.98
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dichloro-5-(trifluoromethyl)pyridine
EINECSNumber (5)-5698
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.