Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-Dichloroacetophenone, 98%

Catalog No. AAA1266706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1266706 5 g
AAA1266703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1266706 Supplier Thermo Scientific Chemicals Supplier No. A1266706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56041-57-7 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56041-57-7
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00052988
InChI Key KMABBMYSEVZARZ-UHFFFAOYSA-N
Synonym 1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
PubChem CID 91986
IUPAC Name 1-(2,3-dichlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC(Cl)=C1Cl

Specifications

Density 1.293
Boiling Point 106°C to 108°C (2 mmHg)
Refractive Index 1.56
Quantity 5 g
Beilstein 2438815
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',3'-Dichloroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 259-954-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.