Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific™

Catalog No. AAH54858AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54858AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54858AE Supplier Thermo Scientific Chemicals Supplier No. H54858AE

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946491-90-3
Molecular Formula C7H5Cl3Mg
Molecular Weight (g/mol) 219.772
MDL Number MFCD11553211
InChI Key OERXPQYEBNEZIP-UHFFFAOYSA-M
Synonym 2,3-dichlorobenzylmagnesium chloride, 2,3-dichlorobenzylmagnesium chloride, 0.25m in ether, 2,3-dichlorobenzylmagnesium chloride, 0.25m in 2-methf
PubChem CID 73996000
SMILES [CH2-]C1=C(C(=CC=C1)Cl)Cl.[Mg+2].[Cl-]

Specifications

Quantity 100 mL
UN Number UN2924
Formula Weight 219.78
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 2,3-Dichlorobenzylmagnesium chloride
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.