Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobutane 98%, Thermo Scientific™

Catalog No. AC113271000
Change view
Click to view available options
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC113271000 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC113271000 Supplier Thermo Scientific Chemicals Supplier No. 113271000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7581-97-7
CAS Min % 97.5
CAS Max % 100.0
Density 1.1000g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H8Cl2
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Linear Formula CH3CH(Cl)CH(Cl)CH3
Quantity 100mL
MDL Number MFCD00000866
Beilstein 01, 119
Synonym butane, 2,3-dichloro, meso-2,3-dichlorobutane, butane, 2,3-dichloro-, meso, 2,3-dichloro-butane, dsstox_cid_1565, acmc-209p0a, dsstox_rid_76214, dsstox_gsid_21565
InChI Key RMISVOPUIFJTEO-UHFFFAOYSA-N
SMILES CC(C(C)Cl)Cl
IUPAC Name 2,3-dichlorobutane
Molecular Weight (g/mol) 127.008
PubChem CID 24233
Formula Weight 127.01
Percent Purity ≥97.5%
Chemical Name or Material 2, 3-Dichlorobutane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-486-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.