Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobutane, (+/-) + meso, 99%, Thermo Scientific™

Catalog No. AAB2138614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2138614 25 g
AAB2138606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2138614 Supplier Thermo Scientific Chemicals Supplier No. B2138614

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7581-97-7
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.008
MDL Number MFCD00000866
InChI Key RMISVOPUIFJTEO-UHFFFAOYSA-N
Synonym butane, 2,3-dichloro, butane, 2,3-dichloro-, r*,s*, meso-2,3-dichlorobutane, butane, 2,3-dichloro-, meso, 2,3-dichloro-butane, dsstox_cid_1565, acmc-209p0a, dsstox_rid_76214, dsstox_gsid_21565
PubChem CID 24233
IUPAC Name 2,3-dichlorobutane
SMILES CC(C(C)Cl)Cl

Specifications

Melting Point -80°C
Boiling Point 117°C to 119°C
Flash Point 18°C (65°F)
Odor Chloroform-like
Linear Formula CH3CHClCHClCH3
Quantity 25 g
UN Number UN1993
Formula Weight 127.01
Percent Purity 99%
Chemical Name or Material 2,3-Dichlorobutane, (+/-) + meso
Hazard Category Danger
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
P210-P261-P273-P243
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Take precautionary measures against static discharge.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.