Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloronitrobenzene 99.0+%, TCI America™
SDP

Catalog No. D038625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D038625G 25 g
D0386500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D038625G Supplier TCI America Supplier No. D038625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3209-22-1
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00007062
InChI Key CMVQZRLQEOAYSW-UHFFFAOYSA-N
Synonym 2,3-dichloronitrobenzene, 2,3-dichloro-1-nitrobenzene, benzene, 1,2-dichloro-3-nitro, benzene, dichloronitro, unii-985ews35kg, ccris 5901, pubchem10770, acmc-1ccaw, 2,3 dichloronitrobenzene, dichloronitrobenzene
PubChem CID 18555
IUPAC Name 1,2-dichloro-3-nitrobenzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)[N+](=O)[O-]

Specifications

Melting Point 62°C
Boiling Point 257°C
Quantity 25 g
UN Number 2811
Formula Weight 192.00
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloronitrobenzene
EINECSNumber (3)-0455
RTECSNumber CZ5240000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.