Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenol, 98+%

Catalog No. AAB2294336
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 576-24-9 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-24-9
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002160
InChI Key UMPSXRYVXUPCOS-UHFFFAOYSA-N
Synonym phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
PubChem CID 11334
IUPAC Name 2,3-dichlorophenol
SMILES OC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 57°C to 60°C
Boiling Point 206°C
Flash Point 115°C (239°F)
Odor Phenol-like
Quantity 500 g
UN Number UN2020
Beilstein 2043615
Formula Weight 163
Percent Purity ≥98%
Chemical Name or Material 2,3-Dichlorophenol
Hazard Category H302-H314
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 209-399-8
RTECSNumber SK8450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.