Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenyl isocyanate, 97%

Catalog No. AAL1478003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1478003 1 g
AAL1478006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1478003 Supplier Thermo Scientific Chemicals Supplier No. L1478003
Add to Cart

CAS: 41195-90-8 | C7H3Cl2NO | 188.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41195-90-8
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002000
InChI Key FYWJWWMKCARWQG-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl isocyanate, 2,3-dichlorophenylisocyanate, benzene, 1,2-dichloro-3-isocyanato, 1,2-dichloro-3-isocyanato-benzene, 2,3-dichlorobenzenisocyanate, benzene, dichloroisocyanato, dichlorphenylisocyanate, dichlorophenyl isocyanate, acmc-1ap4f, 2,3dichlorophenylisocyanate
PubChem CID 33050
IUPAC Name 1,2-dichloro-3-isocyanatobenzene
SMILES ClC1=CC=CC(N=C=O)=C1Cl

Specifications

Density 1.424
Boiling Point 58°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5784
Quantity 1 g
UN Number UN2206
Beilstein 4669222
Sensitivity Moisture sensitive
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenyl isocyanate
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 255-253-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.