Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenyl isothiocyanate 97%, Thermo Scientific™

Catalog No. SB01755DE
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01755DE 5 g
SB01755EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB01755DE Supplier Thermo Scientific Chemicals Supplier No. SB01755DE
Add to Cart

Chemical Identifiers

CAS 6590-97-2
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00041048
InChI Key RQZIODPVCCTBAQ-UHFFFAOYSA-N
PubChem CID 2796281
IUPAC Name 1,2-dichloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(N=C=S)=C1Cl

Specifications

Density 1.4340g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 2,3-dichlorophenyl isothiocyanate, 2,3-dichlorophenylisothiocyanate, benzene,1,2-dichloro-3-isothiocyanato, benzene, 1,2-dichloro-3-isothiocyanato, dichlorophenylisothiocyanate, dichlorophenyl isothiocyanate, acmc-1b4g1, 2,3-dichlorophenyl-isothiocyanate, 1,2-dichloro-3-isothiocyanato-benzene, 2,3-dichloro-1-isothiocyanato-benzene
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2, 3-Dichlorophenyl isothiocyanate
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.