Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D343425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3434-25G 25 g
D34345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3434-25G Supplier TCI America Supplier No. D343425G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 151169-74-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01075703
InChI Key TYIKXPOMOYDGCS-UHFFFAOYSA-N
PubChem CID 2734661
IUPAC Name (2,3-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=CC=C1

Specifications

Melting Point 125°C
Quantity 25 g
Synonym 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride
Formula Weight 190.81
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.