Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloropyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D50815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5081-5G 5 g
D50811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5081-5G Supplier TCI America Supplier No. D50815G
Add to Cart
Add to Cart

Specifications

CAS 951677-39-7
Melting Point 141°C
Color White
Molecular Formula C5H4BCl2NO2
Quantity 5 g
MDL Number MFCD04038756
Synonym 2,3-dichloropyridine-4-boronic acid, 2,3-dichloro-4-pyridineboronic acid, 2,3-dichloropyridin-4-yl boronic acid, 2,3-dichloropyridine-4-boronicacid, 2,3-dichloro-4-pyridineboronicacid
InChI Key ABRTZUPJNVJMHP-UHFFFAOYSA-N
SMILES B(C1=C(C(=NC=C1)Cl)Cl)(O)O
IUPAC Name (2,3-dichloropyridin-4-yl)boronic acid
Molecular Weight (g/mol) 191.802
PubChem CID 3478955
Formula Weight 191.80
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.