Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorotetrahydrofuran 90.0+%, TCI America™
SDP

Catalog No. D09855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09855G 5 g
D098525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09855G Supplier TCI America Supplier No. D09855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3511-19-1
Molecular Formula C4H6Cl2O
Molecular Weight (g/mol) 140.991
MDL Number MFCD00059736
InChI Key ZQHLMWUFVRLDRK-UHFFFAOYSA-N
Synonym Tetrahydro-2,3-dichlorofuran
PubChem CID 19025
IUPAC Name 2,3-dichlorooxolane
SMILES C1COC(C1Cl)Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 63°C
Formula Weight 140.99
Percent Purity ≥90.0% (GC)
Chemical Name or Material 2,3-Dichlorotetrahydrofuran
RTECSNumber LU0525000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.