Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dicyano-6-methyl-5-phenylpyrazine, 97%, Thermo Scientific™

Catalog No. AAH5876006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5876006 5 g
AAH5876003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5876006 Supplier Thermo Scientific Chemicals Supplier No. H5876006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52109-67-8
Molecular Formula C13H8N4
Molecular Weight (g/mol) 220.24
MDL Number MFCD05863478
InChI Key YKYXESCSPXPMKE-UHFFFAOYSA-N
Synonym 2,3-dicyano-5-methyl-6-phenylpyrazine, 5-methyl-6-phenyl-2,3-pyrazinedicarbonitrile, 2,3-pyrazinedicarbonitrile, 5-methyl-6-phenyl, 2,3-dicyano-6-methyl-5-phenylpyrazine, 5-methyl-6-phenyl-2,3-pyrazinedicarbonitrile #
PubChem CID 142908
IUPAC Name 5-methyl-6-phenylpyrazine-2,3-dicarbonitrile
SMILES CC1=NC(C#N)=C(N=C1C1=CC=CC=C1)C#N

Specifications

Quantity 5 g
UN Number UN3439
Formula Weight 220.23
Percent Purity 97%
Chemical Name or Material 2,3-Dicyano-6-methyl-5-phenylpyrazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.