Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dicyanohydroquinone, 98%

Catalog No. AC232050250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232050250 25 g Glass bottle
AC232050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232050250 Supplier Thermo Scientific Chemicals Supplier No. 232050250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4733-50-0 | C8H4N2O2 | 160.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4733-50-0
MDL Number MFCD00001790
InChI Key MPAIWVOBMLSHQA-UHFFFAOYSA-N
Synonym 2,3-dicyanohydroquinone, 3,6-dihydroxyphthalonitrile, 2,3-dicyano-p-hydroquinone, 3,6-dihydroxyphthalodinitrile, 1,2-benzenedicarbonitrile, 3,6-dihydroxy, 2,3-dicyanobenzene-1,4-diol, phthalonitrile, 3,6-dihydroxy, dicyanohydroquinone, 2,3-dicyanohydroquinme, acmc-1asrw
PubChem CID 78467
IUPAC Name 3,6-dihydroxybenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C(=C1O)C#N)C#N)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >230.0°C
Color Brown to Khaki
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H4N2O2
Linear Formula (NC)2C6H2-1,4-(OH)2
Quantity 25 g
Molecular Weight (g/mol) 160.132
Formula Weight 160.13
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2, 3-Dicyanohydroquinone, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-241-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.