Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dicyanonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D19771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19771G Supplier TCI America Supplier No. D19771G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 22856-30-0
Melting Point 257°C
Color White-Yellow
Molecular Formula C12H6N2
Quantity 1 g
MDL Number MFCD00059063
UN Number 3439
Synonym 2,3-Naphthalenedicarbonitrile
InChI Key KNBYJRSSFXTESR-UHFFFAOYSA-N
SMILES C1=CC=C2C=C(C(=CC2=C1)C#N)C#N
IUPAC Name naphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol) 178.194
PubChem CID 601270
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dicyanonaphthalene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.