Learn More
Medchemexpress LLC 2',3'-dideoxy-5-iodocytidine | 114748-57-1 | 99.3% | 337.11 g·mol⁻¹ | C9H12IN3O3 | 100 MG
SDP
Catalog No. 5000210437
Encompass_Preferred
2',3'-dideoxy-5-iodocytidine is an iodinated nucleoside analog used as a reagent for gene sequencing and as a research antibacterial agent with reported activity against Mycobacterium. It is supplied as a powder in milligram-scale quantities and is intended for laboratory research use.
  • Iodinated nucleoside analog for sequencing applications.
  • Reported antibacterial activity against Mycobacterium.
  • High purity: 99.29%.
  • Molecular weight 337.11 g·mol⁻¹; formula C9H12IN3O3.
  • Available in 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg sizes.
  • Powder storage: -20°C stable for up to 3 years; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-10437 Supplier Medchemexpress LLC Supplier No. HYW048478100MG
2',3'-dideoxy-5-iodocytidine is an iodinated nucleoside analog used as a reagent for gene sequencing and as a research antibacterial agent with reported activity against Mycobacterium. It is supplied as a powder in milligram-scale quantities and is intended for laboratory research use.
  • Iodinated nucleoside analog for sequencing applications.
  • Reported antibacterial activity against Mycobacterium.
  • High purity: 99.29%.
  • Molecular weight 337.11 g·mol⁻¹; formula C9H12IN3O3.
  • Available in 25 mg, 50 mg, 100 mg, 200 mg, and 500 mg sizes.
  • Powder storage: -20°C stable for up to 3 years; in solvent: -80°C for 6 months, -20°C for 1 month.