Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3'-dideoxy-5-iodouridine | 105784-83-6 | MFCD00074877 | 97.3% | 338.10 g·mol⁻¹ | C9H11IN2O4 | 250 MG
SDP

Catalog No. 5000209446
Encompass_Preferred

2',3'-dideoxy-5-iodouridine is a nucleoside analog used for gene sequencing and as a research reagent in antiviral and anticancer studies. It is supplied as a high-purity solid with manufacturer guidance for storage and handling.

  • Used for gene sequencing and research into antiviral and anticancer activity.
  • Nucleoside analog with CAS number 105784-83-6.
  • Molecular weight 338.10 g·mol⁻¹ and formula C9H11IN2O4.
  • High purity (97.3%) suitable for analytical applications.
  • Available in small-scale quantities (e.g., 250 MG) for laboratory use.
  • Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09446 Supplier Medchemexpress LLC Supplier No. 250MGHYW048476
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3'-dideoxy-5-iodouridine is a nucleoside analog used for gene sequencing and as a research reagent in antiviral and anticancer studies. It is supplied as a high-purity solid with manufacturer guidance for storage and handling.

  • Used for gene sequencing and research into antiviral and anticancer activity.
  • Nucleoside analog with CAS number 105784-83-6.
  • Molecular weight 338.10 g·mol⁻¹ and formula C9H11IN2O4.
  • High purity (97.3%) suitable for analytical applications.
  • Available in small-scale quantities (e.g., 250 MG) for laboratory use.
  • Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.