Learn More
Cayman Chemical 2 3-DIdeoxyadenosIn-5-O-2.5mg
SDP
Catalog No. 502916734
Encompass_Preferred
A sulfur-containing nucleoside triphosphate; prevents complete digestion by 5’-phosphodoiesterase, which allows for alkylation and identification of SNP-containing alleles when incorporated into allele-specific primers; has been used for chain extension in PCR assays; a mixture of Rp and Sp isomers
Catalog No. 50-291-6734 Supplier Cayman Chemical Supplier No. 384602.5 MG
A sulfur-containing nucleoside triphosphate; prevents complete digestion by 5’-phosphodoiesterase, which allows for alkylation and identification of SNP-containing alleles when incorporated into allele-specific primers; has been used for chain extension in PCR assays; a mixture of Rp and Sp isomers