Learn More
Cayman Chemical 2′,3′-Dideoxyadenosine, Purity: ≥98%, CAS: 4097-22-7, 50 mg
SDP
Catalog No. 502913590
Encompass_Preferred
A prodrug form of ddATP; inhibits HIV replication in HIV-infected MT-4 cells at 5 and 125 µM; protects ATH8 cells against the cytopathic effect of HIV (EC50 = 2.7 µM)
Catalog No. 50-291-3590 Supplier Cayman Chemical Supplier No. 3666250 MG
A prodrug form of ddATP; inhibits HIV replication in HIV-infected MT-4 cells at 5 and 125 µM; protects ATH8 cells against the cytopathic effect of HIV (EC50 = 2.7 µM)