Learn More
Medchemexpress LLC HY-W013441 10mg Medchemexpress, 2',3'-Dideoxyadenosine CAS:4097-22-7 Purity:>98%
SDP
Catalog No. 501970696
Encompass_Preferred
Medchemexpress, HY-W013441 10mg 2',3'-Dideoxyadenosine CAS:4097-22-7 2',3'-Dideoxyadenosine is an inhibitor of HIV replication. Antiretroviral activity. Antiviral efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0696 Supplier Medchemexpress LLC Supplier No. HYW01344110MG
Medchemexpress, HY-W013441 10mg 2',3'-Dideoxyadenosine CAS:4097-22-7 2',3'-Dideoxyadenosine is an inhibitor of HIV replication. Antiretroviral activity. Antiviral efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.