Learn More
Selleck Chemical LLC 2 3 -Dideoxyadenosine
SDP
Catalog No. 5000004148
Encompass_Preferred
2 3 -Dideoxyadenosine (ddA ddAdo) is an anti-human immunodeficiency virus agent against human immunodeficiency virus (HIV) and human hepatitis B virus (HBV) replication
Catalog No. 50-000-04148 Supplier Selleck Chemical LLC Supplier No. S59795MG
2 3 -Dideoxyadenosine (ddA ddAdo) is an anti-human immunodeficiency virus agent against human immunodeficiency virus (HIV) and human hepatitis B virus (HBV) replication