Learn More
Medchemexpress LLC 2',3'-Dideoxyadenosine 5'-triphosphate (trisodium) | 72029-21-1 | 99.9% | 27 MG
SDP
Catalog No. 5000340948
Encompass_Preferred
The product is ddATP (trisodium) solution (100mM), also known as 2',3'-Dideoxyadenosine 5'-triphosphate (trisodium). This solution serves as an active metabolite of 2',3'-dideoxyinosine and functions as a chain-elongating inhibitor of DNA polymerase. It is applicable in the Sanger method for DNA sequencing and for research concerning virus infections.
  • Chain-elongating inhibitor of DNA polymerase.
  • Utilized in the Sanger method for DNA sequencing.
  • Supports research on virus infection.
  • Inhibits adenovirus DNA synthesis in infected cells.
  • Reduces γH2A.X foci in DNA double-strand breaks (DSB) oocytes.
  • Inhibits HIV replication by targeting HIV reverse transcriptase.
Catalog No. 50-003-40948 Supplier Medchemexpress LLC Supplier No. HY128036B27MG
The product is ddATP (trisodium) solution (100mM), also known as 2',3'-Dideoxyadenosine 5'-triphosphate (trisodium). This solution serves as an active metabolite of 2',3'-dideoxyinosine and functions as a chain-elongating inhibitor of DNA polymerase. It is applicable in the Sanger method for DNA sequencing and for research concerning virus infections.
  • Chain-elongating inhibitor of DNA polymerase.
  • Utilized in the Sanger method for DNA sequencing.
  • Supports research on virus infection.
  • Inhibits adenovirus DNA synthesis in infected cells.
  • Reduces γH2A.X foci in DNA double-strand breaks (DSB) oocytes.
  • Inhibits HIV replication by targeting HIV reverse transcriptase.