Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3'-dideoxycytidine | 7481-89-2 | MFCD00012188 | 98.6% | 211.22 | C9H13N3O3 | 10 MG
SDP

Catalog No. 5000298165
Encompass_Preferred

Zalcitabine (standard) is an analytical reference standard of the nucleoside analogue reverse transcriptase inhibitor zalcitabine (2',3'-dideoxycytidine) intended for research and analytical applications. Supplied as a white to off-white solid, it is used for method development, qualitative and quantitative assays, and instrument calibration in analytical workflows.

  • High purity suitable for analytical assays.
  • Characterized by molecular formula C9H13N3O3 and molecular weight 211.22.
  • Applicable for HPLC, GC, and MS method development and validation.
  • Delivered as a solid in defined small pack sizes for reference use.
  • Provided with documentation for proper storage and handling.

Catalog No. 50-002-98165 Supplier Medchemexpress LLC Supplier No. HY17392R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zalcitabine (standard) is an analytical reference standard of the nucleoside analogue reverse transcriptase inhibitor zalcitabine (2',3'-dideoxycytidine) intended for research and analytical applications. Supplied as a white to off-white solid, it is used for method development, qualitative and quantitative assays, and instrument calibration in analytical workflows.

  • High purity suitable for analytical assays.
  • Characterized by molecular formula C9H13N3O3 and molecular weight 211.22.
  • Applicable for HPLC, GC, and MS method development and validation.
  • Delivered as a solid in defined small pack sizes for reference use.
  • Provided with documentation for proper storage and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.