Learn More
Cayman Chemical 2 3-DIdeoxygunosIn-5-O-2.5mg
SDP
Catalog No. 502916731
Encompass_Preferred
A sulfur-containing nucleoside triphosphate derivative; prevents complete digestion by 5’-phosphodiesterase, which allows for alkylation and identification of SNP-containing alleles when incorporated into allele-specific primers; has been used for chain extension using a mutated polymerase selective for dideoxynucleotides and chain termination in allele-specific PCR assays
Catalog No. 50-291-6731 Supplier Cayman Chemical Supplier No. 384622.5 MG
A sulfur-containing nucleoside triphosphate derivative; prevents complete digestion by 5’-phosphodiesterase, which allows for alkylation and identification of SNP-containing alleles when incorporated into allele-specific primers; has been used for chain extension using a mutated polymerase selective for dideoxynucleotides and chain termination in allele-specific PCR assays