Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3'-dideoxyuridine | 5983-09-5 | MFCD00013359 | 99.9% | 212.21 g·mol⁻1 | C9H12N2O4 | 250 MG
SDP

Catalog No. 5000302702
Encompass_Preferred

2',3'-Dideoxyuridine is a purine nucleoside analog used in biochemical and pharmacological research as a nucleoside analog and building block for studies of DNA synthesis and nucleoside metabolism. It is provided as a white to off-white powder suitable for analytical and research applications.

  • High purity (99.9%) suitable for research use.
  • Molecular formula C9H12N2O4 and molecular weight 212.21 g·mol⁻1.
  • CAS number 5983-09-5 for unambiguous identification.
  • Available in multiple pack sizes for flexibility in experiments.
  • Supplied as a solid powder for easy weighing and storage.

Catalog No. 50-003-02702 Supplier Medchemexpress LLC Supplier No. 250MGHYW014306
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3'-Dideoxyuridine is a purine nucleoside analog used in biochemical and pharmacological research as a nucleoside analog and building block for studies of DNA synthesis and nucleoside metabolism. It is provided as a white to off-white powder suitable for analytical and research applications.

  • High purity (99.9%) suitable for research use.
  • Molecular formula C9H12N2O4 and molecular weight 212.21 g·mol⁻1.
  • CAS number 5983-09-5 for unambiguous identification.
  • Available in multiple pack sizes for flexibility in experiments.
  • Supplied as a solid powder for easy weighing and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.