Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diethylpyrazine, 98%, Thermo Scientific™

Catalog No. AAA1327706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1327706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1327706 Supplier Thermo Scientific Chemicals Supplier No. A1327706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15707-24-1
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00006151
InChI Key GZXXANJCCWGCSV-UHFFFAOYSA-N
Synonym pyrazine, 2,3-diethyl, unii-ad31a42vy0, pyrazine, diethyl, fema no. 3136, 2,3-diethyl-pyrazine, shoyu pyrazine, pubchem8606, 2,3-diethyl pyrazine, acmc-1brc9, dsstox_cid_27101
PubChem CID 27458
IUPAC Name 2,3-diethylpyrazine
SMILES CCC1=NC=CN=C1CC

Specifications

Density 0.963
Boiling Point 180°C to 182°C
Flash Point 71°C (160°F)
Odor Stench
Refractive Index 1.5
Quantity 5 g
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 2,3-Diethylpyrazine
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 239-800-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.