Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorobenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. D26145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26145G 5 g
D261425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26145G Supplier TCI America Supplier No. D26145G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2646-91-5
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010292
InChI Key WDBAXYQUOZDFOJ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3-difluoro, 2,3-difluoro benzaldehyde, 2,3-difluoro-benzaldehyde, difluoro-benzaldehyde, pubchem1436, 2,3 difluorobenzaldehyde, acmc-1cr6r, 2,3-difluorobenzaldehyde?, 2,3-difluorobenzaldehyde, ksc493k8r
PubChem CID 137664
IUPAC Name 2,3-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)F)C=O

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
UN Number 1989
Formula Weight 142.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.