Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorobenzoyl chloride, 98%, Thermo Scientific™

Catalog No. AC375820010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC375820010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC375820010 Supplier Thermo Scientific Chemicals Supplier No. 375820010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 18355-73-2
Density 1.4230g/mL
Boiling Point 85.0°C to 87.0°C (14.0 mmHg)
Flash Point 91°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H3ClF2O
Quantity 1 g
MDL Number MFCD00042499
Specific Gravity 1.423
Synonym 2,3-difluorobenzoylchloride, benzoylchloride, 2,3-difluoro, benzoyl chloride, 2,3-difluoro, pubchem16225, acmc-1cabv, 2,3-difluoro-benzoyl chloride, 2,3-bis fluoranyl benzoyl chloride, 2,3-difluorobenzoyl chloride, 3-chlorocarbonyl-1,2-difluorobenzene
InChI Key CDCFERWURRNLLA-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)F)F)C(=O)Cl
IUPAC Name 2,3-difluorobenzoyl chloride
Molecular Weight (g/mol) 176.55
PubChem CID 519549
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2, 3-Difluorobenzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.