Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorophenetole 98.0+%, TCI America™
SDP

Catalog No. D38885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38885G Supplier TCI America Supplier No. D38885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121219-07-6
Molecular Formula C8H8F2O
Molecular Weight (g/mol) 158.148
MDL Number MFCD07368737
InChI Key AVOGLGBKOFOSBN-UHFFFAOYSA-N
Synonym 1-Ethoxy-2,3-difluorobenzene
PubChem CID 2782928
IUPAC Name 1-ethoxy-2,3-difluorobenzene
SMILES CCOC1=C(C(=CC=C1)F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 158.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorophenetole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.