Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorophenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. D46021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46021G 1 g
D46025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46021G Supplier TCI America Supplier No. D46021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145689-34-5
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00061277
InChI Key IYRCHGRRMKOSHW-UHFFFAOYSA-N
Synonym 2,3-difluorophenylacetonitrile, 2-2,3-difluorophenyl acetonitrile, 2,3-difluorobenzyl cyanide, 2,3-difluorophenylacetanitrile, benzeneacetonitrile, 2,3-difluoro, 2-2,3-difluorophenyl ethanenitrile, 2,3-difluorobenzeneacetonitrile, pubchem1587, acmc-209x0i, ksc183q4t
PubChem CID 518968
IUPAC Name 2-(2,3-difluorophenyl)acetonitrile
SMILES FC1=CC=CC(CC#N)=C1F

Specifications

Color Yellow
Quantity 1 g
UN Number 3276
Formula Weight 153.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorophenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.