Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Dihydro-1,4-benzodioxin-6-sulfonyl fluoride

Catalog No. ALD001481G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD001481G 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD001481G Supplier Sigma Aldrich Supplier No. ALD001481G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Geranyllinalool is a diterpene alcohol, commonly used as a fragrant
  • It imparts insecticidal property to xylophagous insects and pine wood.geranyllinalool has been used in studying its influence on pyocyanin and pseudomonas quinolone signal (pqs) production by pseudomonas aeruginosa
  • It may be used as a starting material in the synthesis of teprenone by reacting with acetylated meldrumαs acid

Specifications

CAS 1368844-00-1
Melting Point 66°C to 71°C
Molecular Formula C8H7FO4S
Linear Formula C8H7FO4S
MDL Number MFCD21885632
Quantity 1 mL
Synonym 2,3-Dihydro-1,4-benzodioxin-6-sulfonyl fluoride
Molecular Weight (g/mol) 218.2
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.