Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-dihydro-1,4-benzodioxin-6-ylmethylamine

Catalog No. CDS014411 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014411 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014411 Supplier Sigma Aldrich Supplier No. CDS014411100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-ethoxy-2-cyclopentenone (3-ethoxy-2-cyclopenten-1-one, 3-ethoxycyclopentenone) is a cyclic α-alkoxy α,α-unsaturated ketone. its manganese(iii) acetate based tandem oxidation has been investigated. its various physical properties such as density, refractive index and boiling point have been reported.3-ethoxy-2-cyclopentenone (3-ethoxy-2-cyclopenten-1-one) may be used as a starting material in the synthesis of the following:
  • Allylic cyclopentenyl alcohols like 2-cyclopentenol, 3-methyl-2-cyclopentenol and 3-methoxymethaoxymethyl-2-cyclopentenol
  • 3-aminocyclopent-2-en-1-one
  • (e)-5-(((2s*,3s*)-3-((benzyloxy)methyl)oxiran-2- yl)methylene)-3-ethoxycyclopent-2-enone, an α,α -epoxy-α,α-unsaturated cyclic enone

Specifications

Molecular Formula C9H11NO2
Linear Formula C9H11NO2
MDL Number MFCD02677684
Quantity 5 g
Molecular Weight (g/mol) 165.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.