Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-1,4-benzodioxin-6-ylmethylamine, Thermo Scientific™

Catalog No. CC01313DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC01313DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC01313DA Supplier Thermo Scientific Chemicals Supplier No. CC01313DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17413-10-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key FUDYRLUSXBRPIA-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxin-6-yl methanamine, 2,3-dihydro-1,4-benzodioxin-6-ylmethylamine, 3,4-ethylenedioxybenzylamine, 1,4-benzodioxin-6-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-methylamine, 6-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-6-yl methanamine, 1,4-benzodioxin-6-methanamine,2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-methanamine, 6-aminomethyl-2,3-dihydro-1,4-benzodioxin
PubChem CID 87102
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-ylmethanamine
SMILES C1COC2=C(O1)C=CC(=C2)CN

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 165.19
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-6-ylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.