Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-1,4-benzodioxine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC01501DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01501DA 1 g
CC01501CB 250 mg
CC01501DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC01501DA Supplier Thermo Scientific Chemicals Supplier No. CC01501DA
Add to Cart

Chemical Identifiers

CAS 3663-80-7
Molecular Formula C9H7O4
Molecular Weight (g/mol) 179.15
MDL Number MFCD00084888
InChI Key HMBHAQMOBKLWRX-QMMMGPOBSA-M
Synonym 1,4-benzodioxan-2-carboxylic acid, 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-2-carboxylic acid, 1,4-benzodioxin-2-carboxylic acid, 2,3-dihydro, 1,4-benzodioxan-2-carboxylicacid, 2rs-2,3-dihydro-1,4-benzodioxine-2-carboxylic acid, rac 1,4-benzodioxane-2-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-2-carboxylic acid
PubChem CID 2735450
IUPAC Name 2,3-dihydro-1,4-benzodioxine-3-carboxylic acid
SMILES [O-]C(=O)[C@@H]1COC2=CC=CC=C2O1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.