Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Dihydro-[1,4]dioxino[2,3-b]pyridin-8-amine

Catalog No. ADE0000261G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0000261G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0000261G Supplier Sigma Aldrich Supplier No. ADE0000261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The following aryl fluorosulfate can be utilized as a cross-coupling partner in suzuki-miyaura reactions
  • The standard reaction conditions for these palladium-catalyzed c-c bond formations are ligand-free and can be performed in water, at room temperatre and are not air sensitive.palladium-catalyzed, ligand-free suzuki reaction in water using aryl fluorosulfates

Specifications

CAS 1246088-52-7
Molecular Formula C7H8N2O2
Linear Formula C7H8N2O2
MDL Number MFCD17171322
Quantity 500 mg
Molecular Weight (g/mol) 152.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.