Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-dihydro-2-[1-(phenylmethyl)-4-piperidinyl]-4H-1-benzothiopyran-4-one | 1204401-49-9 | >=98.0% | 337.48 | C21H23NOS | 5 MG
SDP

Catalog No. 5000243027
Encompass_Preferred

σ1 Receptor antagonist-1 (CAS 1204401-49-9) is a selective sigma-1 receptor antagonist provided for research use. It has been reported to inhibit cell growth, cause G0/G1 cell cycle arrest, and induce apoptosis in certain cancer cell lines. The compound is supplied as a solid and is intended for in vitro pharmacology and receptor biology studies.

  • Selective sigma-1 receptor antagonist.
  • Demonstrated to inhibit cell growth and induce G0/G1 cell cycle arrest and apoptosis in cell assays.
  • High purity (≥98% by HPLC).
  • Molecular weight 337.48 g/mol.
  • Chemical formula C21H23NOS.
  • Solid form, light yellow to light brown.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-43027 Supplier Medchemexpress LLC Supplier No. HY108155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

σ1 Receptor antagonist-1 (CAS 1204401-49-9) is a selective sigma-1 receptor antagonist provided for research use. It has been reported to inhibit cell growth, cause G0/G1 cell cycle arrest, and induce apoptosis in certain cancer cell lines. The compound is supplied as a solid and is intended for in vitro pharmacology and receptor biology studies.

  • Selective sigma-1 receptor antagonist.
  • Demonstrated to inhibit cell growth and induce G0/G1 cell cycle arrest and apoptosis in cell assays.
  • High purity (≥98% by HPLC).
  • Molecular weight 337.48 g/mol.
  • Chemical formula C21H23NOS.
  • Solid form, light yellow to light brown.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.