Learn More
Medchemexpress LLC 2,3-dihydro-6,7-diphenyl-2-thioxo-4(1H)-pteridinone | 14892-97-8 | MFCD02167478, MFCD28718010 | 99.9% | 332.38 g·mol⁻¹ | C18H12N4OS | 1 ML
SDP
Catalog No. 5000242276
Encompass_Preferred
SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks nonhomologous end-joining (NHEJ) and is used to increase homology-directed repair (HDR) efficiency in genome-editing applications. The product is provided as a 10 mM solution in DMSO for research use and is characterized by a defined molecular formula and molecular weight.
Catalog No. 50-002-42276 Supplier Medchemexpress LLC Supplier No. HY10784510MM 1ML
SCR7 pyrazine is a small-molecule inhibitor of DNA ligase IV that blocks nonhomologous end-joining (NHEJ) and is used to increase homology-directed repair (HDR) efficiency in genome-editing applications. The product is provided as a 10 mM solution in DMSO for research use and is characterized by a defined molecular formula and molecular weight.