Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 97%

Catalog No. AAH5960206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5960206 5 g
AAH5960203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5960206 Supplier Thermo Scientific Chemicals Supplier No. H5960206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 55745-70-5 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55745-70-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00068058
InChI Key WEBVDBDZSOJGPB-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-carboxaldehyde, 2,3-dihydrobenzo b furan-5-carbaldehyde, 2,3-dihydrobenzofuran-5-carbaldehyde, 2,3-dihydrobenzo b furan-5-carboxaldehyde, 2,3-dihyro-1-benzofuran-5-carbaldehyde, 2,3-dihydro-benzofuran-5-carbaldehyde, 5-coumarancarboxaldehyde, 2,3-dihydro-5-benzofurancarboxaldehyde, 5-benzofurancarboxaldehyde, 2,3-dihydro, 2,3-dihydro-5-benzofurancarbaldehyde
PubChem CID 735901
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbaldehyde
SMILES O=CC1=CC=C2OCCC2=C1

Specifications

Density 1.177
Boiling Point 88°C to 89°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6
Quantity 5 g
Beilstein 124062
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde
Hazard Category H334
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P285-P304+P341-P342+P311-P501c
EINECSNumber 259-788-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.