Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydrobenzo[b]furan-7-ylamine, 97%, Thermo Scientific™

Catalog No. CC00914CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00914CB 250 mg Amber Glass Bottle
CC00914DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00914CB Supplier Thermo Scientific Chemicals Supplier No. CC00914CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13414-56-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD11109313
InChI Key UHHZGSLXPQGPJL-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-7-amine, 2,3-dihydrobenzo b furan-7-ylamine, 2,3-dihydro-benzofuran-7-ylamine, 7-benzofuranamine, 2,3-dihydro, 7-amino-2,3-dihydrobenzofuran, 2,3-dihydro-7-benzofuranamine
PubChem CID 12805950
IUPAC Name 2,3-dihydro-1-benzofuran-7-amine
SMILES NC1=CC=CC2=C1OCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydrobenzo[b]furan-7-ylamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.