Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydrobenzofuran-5-carboxaldehyde 96.0+%, TCI America™
SDP

Catalog No. D385725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D385725G 25 g
D38575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D385725G Supplier TCI America Supplier No. D385725G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55745-70-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00068058
InChI Key WEBVDBDZSOJGPB-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-carboxaldehyde, 2,3-dihydrobenzo b furan-5-carbaldehyde, 2,3-dihydrobenzofuran-5-carbaldehyde, 2,3-dihydrobenzo b furan-5-carboxaldehyde, 2,3-dihyro-1-benzofuran-5-carbaldehyde, 2,3-dihydro-benzofuran-5-carbaldehyde, 5-coumarancarboxaldehyde, 2,3-dihydro-5-benzofurancarboxaldehyde, 5-benzofurancarboxaldehyde, 2,3-dihydro, 2,3-dihydro-5-benzofurancarbaldehyde
PubChem CID 735901
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbaldehyde
SMILES O=CC1=CC=C2OCCC2=C1

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 g
Formula Weight 148.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dihydrobenzofuran-5-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.