Learn More
Cayman Chemical 2,3-Dihydrobenzofuran, Purity: ≥98%, CAS: 496-16-2, 10 g
SDP
Catalog No. 502942174
Encompass_Preferred
A coumaran scaffold and pharmacophore; cytotoxic to HeLa cells (IC50 = 23.86 µg/ml)
Catalog No. 50-294-2174 Supplier Cayman Chemical Supplier No. 3994510 G
A coumaran scaffold and pharmacophore; cytotoxic to HeLa cells (IC50 = 23.86 µg/ml)