Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Dihydrofuran

Catalog No. 200018100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-001-8100ML 100 mL
20-001-81L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-001-8100ML Supplier Sigma Aldrich Supplier No. 200018100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The enantioselective Heck arylation of 2,3-dihydrofuran with aryl iodides was studied.2,3-Dihydrofuran is a versatile reagent used in lanthanide-catalyzed Diels-Alder reactions with 2-pyrones and in Rh(II)-stabilized cycloadditions with vinylcarbenoids.

Specifications

CAS 1191-99-7
Density 0.927 g/mL (at 25°C (literature))
Boiling Point 54°C to 55°C (lit.)
Molecular Formula C4H6O
Refractive Index n20/D 1.423 (literature)
Linear Formula C4H6O
MDL Number MFCD00003205
Quantity 100 mL
Synonym 2,3-DHF
Molecular Weight (g/mol) 70.09
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.